• USA Home
  • ES5534 - ChromegaChiral Chiral CCO, 10 Micron HPLC Column

EMAIL THIS PAGE TO A FRIEND
ES5534

ChromegaChiral Chiral CCO, 10 Micron HPLC Column

10 μm particle size, L × I.D. 15 cm × 4.6 mm

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, Chiral HPLC Columns, ChromegaChiral CCO 10 Micron Chiral Columns, ChromegaChiral CCO Chiral Columns, ChromegaChiral Columns,
Quality Level   100
mfr. no.   ES Industries, 135351-CCO
application(s)   HPLC: suitable
L × I.D.   15 cm × 4.6 mm
matrix active group   carbamoyl phase
particle size   10 μm
pore size   1000 Å pore size
separation technique   chiral

Description

General description

ChromegaChiral CCO is a polysaccharide coated chiral stationary phase and columns which are produced using a unique production process of coating the proven chiral selector, tris-(3,5-di-methylphenyl) carbamoyl cellulose on high purity, high performance silica. ChromegaChiral CCO columns are available in 3, 5, 10, and 20 micron particle sizes enabling easy scale up from analytical to preparative scale using HPLC or SFC conditions.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ChromegaChiral is a trademark of ES Industries

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?