• USA Home
  • K08970954 - Kromasil® Guard Cartridge Holder

EMAIL THIS PAGE TO A FRIEND
K08970954

Kromasil® Guard Cartridge Holder

for use with 2.1 - 4.6 mm I.D. x 10 mm L guard cartridge

  •  eCl@ss 32110501

  •  NACRES NB.21

Purchase

Properties

packaging   pkg of 1 ea
mfr. no.   Kromasil, KR-HOLDER-SMALL-ID
availability   available only in USA, Canada and Puerto Rico
application(s)   HPLC: suitable
Featured Industry   Food and Beverages
compatibility   for use with 2.1 - 4.6 mm I.D. x 10 mm L guard cartridge

Description

Legal Information

Kromasil is a registered trademark of EKA Chemicals AB

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?