• USA Home
  • K08971107 - Kromasil® CelluCoat® Chiral HPLC Guard Cartridge

EMAIL THIS PAGE TO A FRIEND
K08971107

Kromasil® CelluCoat® Chiral HPLC Guard Cartridge

use to protect 10 mm I.D. columns, pkg of 3 ea

  •  eCl@ss 32110501

  •  NACRES NB.21

Purchase

Properties

Related Categories Analytical/Chromatography, CelluCoat, Chiral HPLC Columns, Chiral HPLC Guard Columns and Accessories, HPLC Columns, by Brand for Small Molecules,
packaging   pkg of 3 ea
mfr. no.   Kromasil, KRWP-5-CelluCoat-10XGC
application(s)   HPLC: suitable
L × I.D.   10 mm × 10 mm
matrix active group   carbamoyl phase
particle size   5 μm
pore size   >1000 Å
suitability   suitable for L40 per USP
compatibility   use to protect 10 mm I.D. columns
separation technique   chiral

Description

Legal Information

CelluCoat is a registered trademark of EKA Chemicals AB

Kromasil is a registered trademark of EKA Chemicals AB

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?