• USA Home
  • K08971223 - Kromasil® CelluCoat® Chiral HPLC Column
EMAIL THIS PAGE TO A FRIEND
K08971223
Kromasil® CelluCoat® Chiral HPLC Column
3 μm particle size, L × I.D. 150 mm × 2.1 mm
  •  eCl@ss 32110501
  •  NACRES SB.52
Purchase
Properties
mfr. no.   Kromasil, KRWP-3-CelluCoat-2.1X150
availability   available only in USA, Canada and Puerto Rico
parameter   0-40 °C temperature
  50 bar pressure (725 psi)
application(s)   HPLC: suitable
L × I.D.   150 mm × 2.1 mm
particle size   3 μm
pore size   >1000 Å
suitability   suitable for L40 per USP
separation technique   chiral
Description
Legal Information
CelluCoat is a registered trademark of EKA Chemicals AB
Kromasil is a registered trademark of EKA Chemicals AB
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?