• USA Home
  • MET669A - 2-Chloro-4-nitrophenol

EMAIL THIS PAGE TO A FRIEND
MET669A Supelco

2-Chloro-4-nitrophenol

analytical standard

Purchase

Properties

grade   analytical standard
InChI Key   BOFRXDMCQRTGII-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., MET-669A
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   105-106 °C (lit.)
Featured Industry   Agriculture
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
SK5075000

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?