EMAIL THIS PAGE TO A FRIEND
N11064

Ametryn

analytical standard

Synonym: 2-Ethylamino-4-isopropylamino-6-methylthio-1,3,5-triazine

  • CAS Number 834-12-8

  • Empirical Formula (Hill Notation) C9H17N5S

  • Molecular Weight 227.33

  •  Beilstein/REAXYS Number 613099

  •  EC Number 212-634-7

  •  MDL number MFCD00055398

  •  PubChem Substance ID 329818488

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 500 mg
mfr. no.   Chem Service, Inc., PS-383
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
SMILES string   CCNc1nc(NC(C)C)nc(SC)n1
InChI   1S/C9H17N5S/c1-5-10-7-12-8(11-6(2)3)14-9(13-7)15-4/h6H,5H2,1-4H3,(H2,10,11,12,13,14)
InChI key   RQVYBGPQFYCBGX-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?