EMAIL THIS PAGE TO A FRIEND
N11455

Chlortoluron

analytical standard

Synonym: 3-(3-Chloro-4-methyl)-1,1-dimethylurea

  • CAS Number 15545-48-9

  • Empirical Formula (Hill Notation) C10H13ClN2O

  • Molecular Weight 212.68

  •  Beilstein/REAXYS Number 2647688

  •  EC Number 239-592-2

  •  MDL number MFCD00018275

  •  PubChem Substance ID 329818505

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Food and Cosmetic Component Standards, Standards for Personal Care and Cosmetic Products, neats,
Quality Level   100
grade   analytical standard
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-2078
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Cleaning Products
Cosmetics
Food and Beverages
Personal Care
format   neat
SMILES string   CN(C)C(=O)Nc1ccc(C)c(Cl)c1
InChI   1S/C10H13ClN2O/c1-7-4-5-8(6-9(7)11)12-10(14)13(2)3/h4-6H,1-3H3,(H,12,14)
InChI key   JXCGFZXSOMJFOA-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?