EMAIL THIS PAGE TO A FRIEND
N11877 Supelco

Ethoxyquin

analytical standard

Synonym: 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline

  • CAS Number 91-53-2

  • Empirical Formula (Hill Notation) C14H19NO

  • Molecular Weight 217.31

  •  Beilstein Registry Number 158223

  •  EC Number 202-075-7

  •  MDL number MFCD00023883

Purchase

Properties

grade   analytical standard
InChI Key   DECIPOUIJURFOJ-UHFFFAOYSA-N
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-432
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
density   1.03 g/mL at 20 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
WGK Germany 
1
RTECS 
VB8225000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replacement part

Product #

Description

Add to Cart

31519 Ethoxyquin, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?