EMAIL THIS PAGE TO A FRIEND
N12279 Supelco

Isoproturon

analytical standard

Synonym: 3-(4-Isopropylphenyl)-1,1-dimethylurea

  • CAS Number 34123-59-6

  • Empirical Formula (Hill Notation) C12H18N2O

  • Molecular Weight 206.28

  •  Beilstein/REAXYS Number 2214033

  •  EC Number 251-835-4

  •  MDL number MFCD00078684

  •  PubChem Substance ID 329818528

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Food and Cosmetic Component Standards, Standards for Personal Care and Cosmetic Products, neats,
grade   analytical standard
InChI Key   PUIYMUZLKQOUOZ-UHFFFAOYSA-N
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-2000
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of Atrazine Herbicides on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Atrazine Herbicides on Ascentis® Express C18
Keywords: Chromatography, High performance liquid chromatography, Pesticides, Separation

HPLC Analysis of Phenyl Urea Pesticides on Ascentis® Express C18 Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Phenyl Urea Pesticides on Ascentis® Express C18 Phase
Keywords: Chromatography, Herbicides, High performance liquid chromatography, Pesticides, Separation, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?