• USA Home
  • N12382 - Metam sodium hydrate

EMAIL THIS PAGE TO A FRIEND
N12382 Supelco

Metam sodium hydrate

analytical standard

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-221
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with acids liberates toxic gas.
RIDADR 
UN2811 - class 6.1 - PG 3 - MP - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?