EMAIL THIS PAGE TO A FRIEND
N13037

Mevinphos

analytical standard

  • CAS Number 7786-34-7

  • Empirical Formula (Hill Notation) C7H13O6P

  • Molecular Weight 224.15

  •  Beilstein/REAXYS Number 1793349

  •  EC Number 232-095-1

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Organophosphate, Pesticide Class, Pesticides & Metabolites Standards,
grade   analytical standard
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., N-13037-250MG
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat
SMILES string   COC(=O)C=C(C)OP(=O)(OC)OC
InChI   1S/C7H13O6P/c1-6(5-7(8)10-2)13-14(9,11-3)12-4/h5H,1-4H3/b6-5+
InChI key   GEPDYQSQVLXLEU-AATRIKPKSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN3278 - class 6.1 - PG 1 - EHS -SMP - Organophosphorus compound, toxic, liquid, n.o.s., HI: all (not BR)
WGK Germany 
3
RTECS 
GQ5250000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, MS Detection

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, MS Detection
Keywords: Chromatography, Gas chromatography, Pesticides, Purification, Size-exclusion chromatography

US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, NPD Detection

From our library of Articles, Sigma-Aldrich presents US EPA Method 8141: GC Analysis of Organophosphorus Pesticides (OP-Pest) on Equity®-5, NPD Detection
Keywords: Chromatography, Gas chromatography, Nitrogen phosphorus detector, Pesticides, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?