• USA Home
  • PHL84057 - Methylophiopogonanone B

EMAIL THIS PAGE TO A FRIEND
PHL84057

Methylophiopogonanone B

New

phyproof® Reference Substance

Synonym: (3R)-2,3-Dihydro-5,7-dihydroxy-3-[(4-methoxyphenyl)methyl]-6,8-dimethyl-4H-1-benzopyran-4-one

  • CAS Number 74805-91-7

  • Empirical Formula (Hill Notation) C19H20O5

  • Molecular Weight 328.36

  •  MDL number MFCD20487835

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Flavonoids, PhytoLab Standards, Phytochemical Standards More...
product line   phyproof® Reference Substance
assay   ≥90.0% (HPLC)
mfr. no.   PhytoLab
storage temp.   2-8°C
SMILES string   O=C1[C@H](CC2=CC=C(OC)C=C2)COC3=C(C)C(O)=C(C)C(O)=C31
InChI   1S/C19H20O5/c1-10-16(20)11(2)19-15(17(10)21)18(22)13(9-24-19)8-12-4-6-14(23-3)7-5-12/h4-7,13,20-21H,8-9H2,1-3H3/t13-/m1/s1
InChI key   UFMAZRUMVFVHLY-CYBMUJFWSA-N

Description

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Safety & Documentation

Safety Information

WGK Germany 
WGK 3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?