EMAIL THIS PAGE TO A FRIEND
PS1054 Supelco

Aldicarb-sulfoxide

analytical standard

  • CAS Number 1646-87-3

  • Empirical Formula (Hill Notation) C7H14N2O3S

  • Molecular Weight 206.26

  •  Beilstein/REAXYS Number 2212914

  •  MDL number MFCD00143566

  •  PubChem Substance ID 24899100

Purchase

Properties

grade   analytical standard
packaging   ampule of 10 mg
mfr. no.   Chem Service, Inc., PS-1054
Featured Industry   Agriculture
SMILES string   CNC(ON=CC(C)(C)S(C)=O)=O
InChI   1S/C7H14N2O3S/c1-7(2,13(4)11)5-9-12-6(10)8-3/h5H,1-4H3,(H,8,10)
InChI key   BXPMAGSOWXBZHS-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2811 - class 6.1 - PG 1 - EHS - Toxic solids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
UE2075000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?