EMAIL THIS PAGE TO A FRIEND
PS105 Supelco

Sodium fluoroacetate

analytical standard

Synonym: Fluoroacetic acid sodium salt, Gifblaar poison

  • CAS Number 62-74-8

  • Linear Formula FCH2COONa

  • Molecular Weight 100.02

  •  Beilstein Registry Number 3915223

  •  EC Number 200-548-2

  •  MDL number MFCD00002682

  •  PubChem Substance ID 24899097

Purchase

Properties

grade   analytical standard
InChI Key   JGFYQVQAXANWJU-UHFFFAOYSA-M
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-105
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2629 6.1 / PGI
WGK Germany 
3
RTECS 
AH9100000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?