EMAIL THIS PAGE TO A FRIEND
PS2014

Triforin

analytical standard

  • CAS Number 26644-46-2

  • Empirical Formula (Hill Notation) C10H14Cl6N4O2

  • Molecular Weight 434.96

  •  Beilstein/REAXYS Number 626358

  •  EC Number 247-872-0

  •  MDL number MFCD00055521

  •  PubChem Substance ID 24899132

Purchase

Properties

grade   analytical standard
form   neat
packaging   ampule of 100 mg
mfr. no.   Chem Service, Inc., PS-2014
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
Environmental
format   neat
SMILES string   ClC(Cl)(Cl)C(NC=O)N1CCN(CC1)C(NC=O)C(Cl)(Cl)Cl
InChI   1S/C10H14Cl6N4O2/c11-9(12,13)7(17-5-21)19-1-2-20(4-3-19)8(18-6-22)10(14,15)16/h5-8H,1-4H2,(H,17,21)(H,18,22)
InChI key   RROQIUMZODEXOR-UHFFFAOYSA-N

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
TK9200000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?