EMAIL THIS PAGE TO A FRIEND
PS2031 Supelco

Formaldehyde

~37 wt. % in H2O, pkg of 1000 mg

Purchase

Properties

packaging   pkg of 1000 mg
mfr. no.   Chem Service, Inc., PS-2031
concentration   ~37 wt. % in H2O
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 2209 8 / PGIII
WGK Germany 
3
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?