EMAIL THIS PAGE TO A FRIEND
PS2033 Supelco

Dicyclopentadiene

analytical standard

Synonym: 4,7-Methano-3a,4,7,7a-tetrahydroindene, Cyclopentadiene dimer, DCPD

  • CAS Number 77-73-6

  • Empirical Formula (Hill Notation) C10H12

  • Molecular Weight 132.20

  •  Beilstein Registry Number 1904092

  •  EC Number 201-052-9

  •  MDL number MFCD00078246

  •  PubChem Substance ID 24899138

Purchase

Properties

grade   analytical standard
InChI Key   HECLRDQVFMWTQS-AFWXGSBKSA-N
form   neat
packaging   ampule of 500 mg
mfr. no.   Chem Service, Inc., PS-2033
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   170 °C(lit.)
mp   33 °C (lit.)
density   0.986 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 2048 3 / PGIII
WGK Germany 
2
Flash Point(F) 
75.2 °F
Flash Point(C) 
24 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?