EMAIL THIS PAGE TO A FRIEND
PS2078

Chlortoluron

analytical standard

Synonym: 3-(3-Chloro-4-methyl)-1,1-dimethylurea

  • CAS Number 15545-48-9

  • Empirical Formula (Hill Notation) C10H13ClN2O

  • Molecular Weight 212.68

  •  Beilstein/REAXYS Number 2647688

  •  EC Number 239-592-2

  •  MDL number MFCD00018275

Purchase

Properties

grade   analytical standard
packaging   pkg of 250 mg
mfr. no.   Chem Service, Inc., PS-2078
Featured Industry   Agriculture
Cleaning Products
Cosmetics
Environmental
Food and Beverages
Personal Care
SMILES string   CN(C)C(=O)Nc1ccc(C)c(Cl)c1
InChI   1S/C10H13ClN2O/c1-7-4-5-8(6-9(7)11)12-10(14)13(2)3/h4-6H,1-3H3,(H,12,14)
InChI key   JXCGFZXSOMJFOA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
YS7230000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?