EMAIL THIS PAGE TO A FRIEND
PS2086 Supelco

Imidacloprid

analytical standard

Purchase

Properties

grade   analytical standard
InChI Key   YWTYJOPNNQFBPC-UHFFFAOYSA-N
form   neat
packaging   ampule of 500 mg
mfr. no.   Chem Service, Inc., PS-2086
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
Gene Information   rat ... Chrna2(170945), Chrna4(25590)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
NJ0560000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?