EMAIL THIS PAGE TO A FRIEND
PS274 Supelco

Picloram

analytical standard

Synonym: 4-Amino-3,5,6-trichloropicolinic acid, 4-Amino-3,5,6-trichloropyridine-2-carboxylic acid, Picloram

  • CAS Number 1918-02-1

  • Empirical Formula (Hill Notation) C6H3Cl3N2O2

  • Molecular Weight 241.46

  •  Beilstein Registry Number 479075

  •  EC Number 217-636-1

  •  MDL number MFCD00012101

  •  PubChem Substance ID 24899175

Purchase

Properties

grade   analytical standard
InChI Key   NQQVFXUMIDALNH-UHFFFAOYSA-N
form   neat
packaging   ampule of 250 mg
mfr. no.   Chem Service, Inc., PS-274
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   200 °C (dec.) (lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
2
RTECS 
TJ7525000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?