• USA Home
  • PS337 - 2,4-D dimethylamine salt

EMAIL THIS PAGE TO A FRIEND
PS337 Supelco

2,4-D dimethylamine salt

analytical standard

Purchase

Properties

grade   analytical standard
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-337
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2
RTECS 
AG8400000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?