EMAIL THIS PAGE TO A FRIEND
PS551

Baygon®

analytical standard

Synonym: Propoxur

  • CAS Number 114-26-1

  • Empirical Formula (Hill Notation) C11H15NO3

  • Molecular Weight 209.24

  •  Beilstein/REAXYS Number 1879891

  •  EC Number 204-043-8

  •  MDL number MFCD00055455

  •  PubChem Substance ID 24899241

Purchase

Properties

grade   analytical standard
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-551
Featured Industry   Agriculture
Environmental
SMILES string   CNC(=O)Oc1ccccc1OC(C)C
InChI   1S/C11H15NO3/c1-8(2)14-9-6-4-5-7-10(9)15-11(13)12-3/h4-8H,1-3H3,(H,12,13)
InChI key   ISRUGXGCCGIOQO-UHFFFAOYSA-N

Description

Legal Information

Baygon is a registered trademark of S. C. Johnson & Son, Inc.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2811 - class 6.1 - PG 2 - MP - RQ - Toxic solids, organic, n.o.s. HI: all
WGK Germany 
3
RTECS 
FC3150000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?