• USA Home
  • PS61 - 1,1-Dimethyl-3-phenylurea

EMAIL THIS PAGE TO A FRIEND
PS61 Supelco

1,1-Dimethyl-3-phenylurea

analytical standard

Synonym: Fenuron

  • CAS Number 101-42-8

  • Empirical Formula (Hill Notation) C9H12N2O

  • Molecular Weight 164.20

  •  Beilstein/REAXYS Number 2208535

  •  EC Number 202-941-4

  •  MDL number MFCD00025662

  •  PubChem Substance ID 24899253

Purchase

Properties

Related Categories A-D, All Herbicide Standards, Analytical Standards, Analytical/Chromatography, Herbicides,
grade   analytical standard
form   neat
packaging   ampule of 1000 mg
mfr. no.   Chem Service, Inc., PS-61
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
Featured Industry   Agriculture
format   neat
SMILES string   CN(C)C(=O)Nc1ccccc1
InChI   1S/C9H12N2O/c1-11(2)9(12)10-8-6-4-3-5-7-8/h3-7H,1-2H3,(H,10,12)
InChI key   XXOYNJXVWVNOOJ-UHFFFAOYSA-N
Gene Information   human ... EPHX2(2053)
mouse ... Ephx2(13850)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
YT1450000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Carbamate and Urea Pesticides on SUPELCOSIL™ LC-8
Keywords: Chromatography, Environmental, Environmental Analysis, Herbicides, High performance liquid chromatography, Pesticides

HPLC Analysis of Phenyl Urea Pesticides on Ascentis® Express C18 Phase

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Phenyl Urea Pesticides on Ascentis® Express C18 Phase
Keywords: Chromatography, Herbicides, High performance liquid chromatography, Pesticides, Separation, Size-exclusion chromatography

HPLC Analysis of Urea Pesticides on Discovery® Cyano

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Urea Pesticides on Discovery® Cyano
Keywords: Chromatography, Environmental, Environmental Analysis, Herbicides, High performance liquid chromatography, Metabolites, Pesticides

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?