• USA Home
  • RAD1221 - radiello Vertical Adapter
EMAIL THIS PAGE TO A FRIEND
RAD1221
radiello Vertical Adapter
non-threaded for ready-to-use samplers, pk of 20
  •  NACRES NB.24
Purchase
Properties
Related Categories radiello® Diffusive Sampling Products, Air Monitoring, Analytical/Chromatography, Diffusive Sampling Accessories, Passive/Diffusive Sampling,
packaging   pk of 20
mfr. no.   Radiello, Code 122-1
application(s)   air monitoring: suitable
Featured Industry   Environmental
Industrial Hygiene
Description
General description
For personal sampling (breathing zone assessment) a vertical adapter is available to position the Radiello diffusive body vertically on the Radiello triangular support plate.
Legal Information
radiello is a trademark of Institi Clinici Scientifici Maugeri
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Articles
How Does the Diffusive Sampler Work and Why is it so Special?
The diffusive sampler is a closed box, usually cylindrical. Of its two opposite sides, one is “transparent” to gaseous molecules which cross it, and are adsorbed onto the second side. The former side...
Keywords: Adsorption, Diffusion, Diffusive sampling, Environmental, Flame ionization detector, Mass spectrometry, PAGE, Sample preparations
How to use the radiello® Diffusive Air Sampler
From our library of Articles, Sigma-Aldrich presents How to use the radiello® Diffusive Air Sampler
What is radiello® diffusive sampling?
In the mid 1990's, Dr. Vincenzo Cocheo, director of the Fondazione Salvatore Maugeri, Padova, Italy, in collaboration with the European Commission's Joint Research Center and other institutions, deve...
Keywords: Diffusion, Gas chromatography, Mass spectrometry, Sample preparations
radiello® Air Sampler Components
The essential parts of radiello are the adsorbing cartridge, the diffusive body, the supporting plate and the adhesive label with the bar code indication. Apart from the adsorbing cartridge, if not d...
Keywords: Sample preparations
radiello® Diffusive Air Sampler Calibration Solutions & Kits
RAD171 relieves you from the task of preparing the sodium sulfide standard solution for the calibration curve used for the determination of H2S by the cartridge RAD170. Since sodium sulfide is deliqu...
Keywords: High performance liquid chromatography, PAGE, Purification, Titrations
radiello® Diffusive Air Sampler Maintenance
When exposed outdoors or in a workplace environment, the diffusive body may get dirty from airborne dust. Fine particles (PM10) are especially harmful to yellow diffusive bodies since they can obstru...
Keywords: Detergents, Solvents
radiello® Diffusive Air Sampling Applications - Aldehydes
Blue diffusive body Product No. RAD1201 Supporting plate Product No. RAD121 Vertical adapter Product No. RAD122 (optional) Chemiadsorbing cartridge Product No. RAD165 Filtration kit Product No. RAD17...
Keywords: Filtration, Gene expression, High performance liquid chromatography, Ozonolysis, PAGE, Sample preparations, Spectroscopy
radiello® Passive Air Sampler Overview and Applications
From our library of Articles, Sigma-Aldrich presents radiello® Passive Air Sampler Overview and Applications
Keywords: Diffusive sampling, Sample preparations
radiello® Ready-to-Use
The ready-to-use version may be advantageous when you prefer not to assemble all of the components on field. It can be purchased as it is or in separate parts to be assembled by the customer. In the ...
Keywords: Sample preparations
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?