• USA Home
  • RAD1232 - radiello ready-to-use Diffusive Sampler
EMAIL THIS PAGE TO A FRIEND
RAD1232
radiello ready-to-use Diffusive Sampler
for sampling BTEX and VOCs (Thermal Desorption), pk of 5
  •  NACRES NB.24
Purchase
Properties
Related Categories radiello® Diffusive Sampling Products, Air Monitoring, Analytical/Chromatography, Passive/Diffusive Sampling, Ready-to-use Diffusive Sampler,
Quality Level   100
description   Includes RAD1202 Yellow Diffusive Body and RAD145 Cartridge Adsorbent
packaging   pk of 5
Featured Industry   Environmental
Industrial Hygiene
compatibility   for sampling BTEX and VOCs (Thermal Desorption)
Description
General description
The Radiello ready-to-use (rtu) Diffusive Samplers come pre-assembled with the cartridge adsorbent pre-sealed within the diffusive body using a polycarbonate screw-thread cap. To avoid premature sampling, the entire unit (diffusive body plus pre-sealed cartridge adsorbent) is enclosed within an airtight polypropylene (PP) container. Just before use, the unit is removed from the PP container, and snapped into the rtu vertical adapter pre-fixed to the triangular support plate. Once sampling is complete, the diffusive sampling unit is removed from the support plate and resealed into the PP container.
Each rtu sampler includes: a sampler unit (sealed diffusive body with cartridge adsorbent), glass or plastic tube to house the cartridge adsorbent after sampling prior to desorption, rtu vertical adapter, barcode label, and PP container. Triangular support plates must be ordered separately.
Note that the rtu diffusive samplers are ideal for work place sampling campaigns, but not for sampling low concentrations in outdoor or domestic environments. Shelf life for the rtu samplers are 3 months.
Legal Information
radiello is a trademark of Institi Clinici Scientifici Maugeri
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Articles
How Does the Diffusive Sampler Work and Why is it so Special?
The diffusive sampler is a closed box, usually cylindrical. Of its two opposite sides, one is “transparent” to gaseous molecules which cross it, and are adsorbed onto the second side. The former side...
Keywords: Adsorption, Diffusion, Diffusive sampling, Environmental, Flame ionization detector, Mass spectrometry, PAGE, Sample preparations
How to use the radiello® Diffusive Air Sampler
From our library of Articles, Sigma-Aldrich presents How to use the radiello® Diffusive Air Sampler
What is radiello® diffusive sampling?
In the mid 1990's, Dr. Vincenzo Cocheo, director of the Fondazione Salvatore Maugeri, Padova, Italy, in collaboration with the European Commission's Joint Research Center and other institutions, deve...
Keywords: Diffusion, Gas chromatography, Mass spectrometry, Sample preparations
radiello® Air Sampler Components
The essential parts of radiello are the adsorbing cartridge, the diffusive body, the supporting plate and the adhesive label with the bar code indication. Apart from the adsorbing cartridge, if not d...
Keywords: Sample preparations
radiello® Diffusive Air Sampler Calibration Solutions & Kits
RAD171 relieves you from the task of preparing the sodium sulfide standard solution for the calibration curve used for the determination of H2S by the cartridge RAD170. Since sodium sulfide is deliqu...
Keywords: High performance liquid chromatography, PAGE, Purification, Titrations
radiello® Diffusive Air Sampler Maintenance
When exposed outdoors or in a workplace environment, the diffusive body may get dirty from airborne dust. Fine particles (PM10) are especially harmful to yellow diffusive bodies since they can obstru...
Keywords: Detergents, Solvents
radiello® Diffusive Air Sampling Application - VOCs Thermally Desorbed
Yellow diffusive body Product No. RAD1202 Supporting plate Product No. RAD121 Vertical adapter Product No. RAD122 (optional) Adsorbing cartridge Product No. RAD145
Keywords: Adsorption, Air sampling, Chromatography, Degradations, Diffusive sampling, Flame ionization detector, Gas chromatography, Gas chromatography mass spectrometry, Gene expression, Mass selective detector, Mass spectrometry, Phase transitions, Sample preparations, Size-exclusion chromatography, Solvents
radiello® Passive Air Sampler Overview and Applications
From our library of Articles, Sigma-Aldrich presents radiello® Passive Air Sampler Overview and Applications
Keywords: Diffusive sampling, Sample preparations
radiello® Ready-to-Use
The ready-to-use version may be advantageous when you prefer not to assemble all of the components on field. It can be purchased as it is or in separate parts to be assembled by the customer. In the ...
Keywords: Sample preparations
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?