• USA Home
  • RAD1261 - radiello On-Field Thermometer
EMAIL THIS PAGE TO A FRIEND
RAD1261 Supelco
radiello On-Field Thermometer
configured for ready-to-use (rtu) samplers, Include one rtu vertical adapter for each thermometer, pk of 3
  •  NACRES NB.24
Purchase
Properties
Related Categories radiello® Diffusive Sampling Products, Air Monitoring, Analytical/Chromatography, Diffusive Sampling Accessories, Passive/Diffusive Sampling,
packaging   pk of 3
mfr. no.   Radiello, Code 126-1
Featured Industry   Environmental
Industrial Hygiene
compatibility   configured for ready-to-use (rtu) samplers
Description
General description
Uptake rates are dependent on temperature; therefore, concentration values obrtained during samplng will be more accurate if precise temperatures are recorded during sampling. Note that temperature variations of up to 4-5 °C can be observed from one area of an urban population to another.
The Radiello On-Field Thermometer acts as a mini (< 1 cm3) temperature measurement station that can be mounted on the triangular support plate in conjunction with the diffusive body via a pre-attached vertical adapter. The thermomemter offers a precision of ± 5 °C between -20 to 80 °C, and can log up to 2048 temperature data points. This allows the researcher to record one temperature reading every 15 min. for 22 days, every 30 min. for 43 days, or 60 minutes for 85 days. The thermometer requires no batteries, and is amemable to harsh weather conditions.
The Radiello On-Field Thermometer Reader (purchased separately using RAD127) connects the thermometer to your PC (via RS232 serial port) allowing the user to program the thermometer before sampling and down load temperature readings after sampling. Each Thermometer has a unique serial number for easy identificatioin. The software package that performs statistical and graphic analysis is NOT included with the reader. The most recent version can be directly purchased vial the manufacture′s website: www.acrsystems.com
Legal Information
radiello is a trademark of Institi Clinici Scientifici Maugeri
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Articles
How Does the Diffusive Sampler Work and Why is it so Special?
The diffusive sampler is a closed box, usually cylindrical. Of its two opposite sides, one is “transparent” to gaseous molecules which cross it, and are adsorbed onto the second side. The former side...
Keywords: Adsorption, Diffusion, Diffusive sampling, Environmental, Flame ionization detector, Mass spectrometry, PAGE, Sample preparations
How to use the radiello® Diffusive Air Sampler
From our library of Articles, Sigma-Aldrich presents How to use the radiello® Diffusive Air Sampler
What is radiello® diffusive sampling?
In the mid 1990's, Dr. Vincenzo Cocheo, director of the Fondazione Salvatore Maugeri, Padova, Italy, in collaboration with the European Commission's Joint Research Center and other institutions, deve...
Keywords: Diffusion, Gas chromatography, Mass spectrometry, Sample preparations
radiello® Air Sampler Accessories & Spare Parts
The diffusive body can be fitted to the supporting plate either in a vertical or horizontal position, the vertical one being more comfortable when radiello is used for personal sampling.
Keywords: Chromatography, Filtration, High performance liquid chromatography, Ion-exchange chromatography, Sample preparations
radiello® Air Sampler Components
The essential parts of radiello are the adsorbing cartridge, the diffusive body, the supporting plate and the adhesive label with the bar code indication. Apart from the adsorbing cartridge, if not d...
Keywords: Sample preparations
radiello® Diffusive Air Sampler Calibration Solutions & Kits
RAD171 relieves you from the task of preparing the sodium sulfide standard solution for the calibration curve used for the determination of H2S by the cartridge RAD170. Since sodium sulfide is deliqu...
Keywords: High performance liquid chromatography, PAGE, Purification, Titrations
radiello® Diffusive Air Sampler Maintenance
When exposed outdoors or in a workplace environment, the diffusive body may get dirty from airborne dust. Fine particles (PM10) are especially harmful to yellow diffusive bodies since they can obstru...
Keywords: Detergents, Solvents
radiello® Passive Air Sampler Overview and Applications
From our library of Articles, Sigma-Aldrich presents radiello® Passive Air Sampler Overview and Applications
Keywords: Diffusive sampling, Sample preparations
radiello® Ready-to-Use
The ready-to-use version may be advantageous when you prefer not to assemble all of the components on field. It can be purchased as it is or in separate parts to be assembled by the customer. In the ...
Keywords: Sample preparations
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?