• USA Home
  • SBSIT10 - Low Level Sulfur Set (dibutyl sulfide)

EMAIL THIS PAGE TO A FRIEND
SBSIT10 Supelco

Low Level Sulfur Set (dibutyl sulfide)

analytical standard, 0-0.1000 wt. % in isooctane:toluene (9:1)

Purchase

Properties

Related Categories ASTM D6334, ASTM D6445, ASTM Petroleum Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   set of 10 × 100 mL
concentration   0-0.1000 wt. % in isooctane:toluene (9:1)
application(s)   AAS: suitable
  ICP: suitable
Agency/Method   ASTM® D6334
  ASTM® D6445
Featured Industry   Petroleum
format   matrix material
storage temp.   room temp

Description

Analysis Note

Concentration Table
Sulfur, Wt.%
• Standard 1 0.0000
• Standard 2 0.0005
• Standard 3 0.0010
• Standard 4 0.0030
• Standard 5 0.0050
• Standard 6 0.0075
• Standard 7 0.0100
• Standard 8 0.0300
• Standard 9 0.0500
• Standard 10 0.1000

General description

Di-n-butyl sulfide in Isooctane-Toluene mixture.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?