• USA Home
  • SU861021 - Super Clean (Base-Plate Design) Gas Purifier
EMAIL THIS PAGE TO A FRIEND
SU861021 Supelco
Super Clean (Base-Plate Design) Gas Purifier
moisture, with indicator
  •  NACRES SB.54
Purchase
Description
General description
Gas purifier helps in enhancing the performance of GC detectors. It can reduce the background signal in flame ionization and nitrogen phosphorus detectors.
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15
References
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?