• USA Home
  • VT000316 - Salmonella enterica subsp. enterica serovar Typhimurium WDCM 00031 Vitroids

EMAIL THIS PAGE TO A FRIEND
VT000316

Salmonella enterica subsp. enterica serovar Typhimurium WDCM 00031 Vitroids

1,000-10,000 CFU mean value range, certified reference material, for microbiology

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Bacteria Standards, Microbiological Standards, Microbiology,
Quality Level   300
grade   certified reference material
  for microbiology
shelf life   limited shelf life, expiry date on the label
storage temp.   −20°C

Description

General description

This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Download your certificate at: http://www.sigma-aldrich.com.

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)

Legal Information

Vitroids is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

WGK Germany 
WGK 3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?