• USA Home
  • VT025046 - Heterotrophic Organisms Vitroids

EMAIL THIS PAGE TO A FRIEND
VT025046 Supelco

Heterotrophic Organisms Vitroids

1000 - 10000 cfu, certified reference material, for microbiology

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Bacteria Standards, Microbiological Standards, Microbiology,
grade   certified reference material
  for microbiology
storage temp.   −20°C

Description

Legal Information

Vitroids is a trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?