• USA Home
  • Z226130 - Spherisorb® SAX HPLC Column

EMAIL THIS PAGE TO A FRIEND
Z226130

Spherisorb® SAX HPLC Column

5 μm particle size, L × I.D. 25 cm × 4.6 mm

  •  eCl@ss 32110501

  •  NACRES SB.52

Purchase

Properties

Related Categories Analytical/Chromatography, Established Column Lines from Other Manufacturers, HPLC Columns, by Brand for Small Molecules, HPLC/UHPLC, SAX,
Quality Level   100
feature   endcapped: no
mfr. no.   Spherisorb
extent of labeling   4% Carbon loading
parameter   20-45 °C temp. range
  400 bar pressure (5801 psi)
application(s)   HPLC: suitable
L × I.D.   25 cm × 4.6 mm
matrix   tetramethyl ammonium
matrix active group   ammonium ion, quaternary phase
particle size   5 μm
pore size   80 Å
surface area   220 cm2
suitability   suitable for L14 per USP
separation technique   anion exchange

Description

General description

Spherisorb® SAX HPLC Column may be resolved by strong anion-exchange chromatography in disaccharide compositional analysis when linked to a Dionex HPLC system.

Strong anion exchange (SAX) HPLC column could be used for analysing phosphopeptide enrichment and fractionation.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Spherisorb is a registered trademark of Waters

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?