• USA Home
  • Z226866 - Supelco® PEEK Fitting
EMAIL THIS PAGE TO A FRIEND
Z226866 Supelco
Supelco® PEEK Fitting
Hex-head nut, long, wrenchtight
Purchase
Description
General description
Biocompatible fittings for all 1/16 in. tubing connections.
Features and Benefits
Two-Piece Wrenchtight Fittings: Have a special double-tapered ferrule which is leak-free to 10,000 psi (700 kg/cm2). Hexagonal head. Double-tapered ferrules included.
Legal Information
Supelco is a registered trademark of Sigma-Aldrich Co. LLC
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis (COA)
Please Enter a Lot Number
Protocols & Articles
Related Content
Supelco HPLC Calculator for Isocratic or Gradient Method Transfer
Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?