EMAIL THIS PAGE TO A FRIEND
1006007 USP

Acetohexamide

United States Pharmacopeia (USP) Reference Standard

Synonym: 4-Acetyl-N-[(cyclohexylamino)carbonyl]benzenesulfonamide

  • CAS Number 968-81-0

  • Empirical Formula (Hill Notation) C15H20N2O4S

  • Molecular Weight 324.40

  •  MDL number MFCD00072156

  •  PubChem Substance ID 329748134

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, USP Standards, USP Standards A - B More...
form   neat
mfr. no.   USP
Featured Industry   Pharmaceutical (small molecule)
format   neat
storage temp.   room temp
SMILES string   CC(=O)c1ccc(cc1)S(=O)(=O)NC(=O)NC2CCCCC2
InChI   1S/C15H20N2O4S/c1-11(18)12-7-9-14(10-8-12)22(20,21)17-15(19)16-13-5-3-2-4-6-13/h7-10,13H,2-6H2,1H3,(H2,16,17,19)
InChI key   VGZSUPCWNCWDAN-UHFFFAOYSA-N
Gene Information   human ... ABCC8(6833), KCNJ11(3767)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Oral hypoglycemic agent

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.

Sales restrictions may apply.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

TLC Analysis of Fatty Acids as Isatinmethyl Esters on HPTLC Silica Gel 60 RP-18 F254s

From our library of Articles, Sigma-Aldrich presents TLC Analysis of Fatty Acids as Isatinmethyl Esters on HPTLC Silica Gel 60 RP-18 F254s
Keywords: Chromatography, Thin layer chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?