EMAIL THIS PAGE TO A FRIEND
1029308 USP

Amlexanox

United States Pharmacopeia (USP) Reference Standard

Synonym: 2-Amino-7-(1-methylethyl)-5-oxo-5H-[1]Benzopyrano[2,3-b]pyridine-3-carboxylic acid, AA 673, Amoxanox, CHX 3673

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, USP Standards, USP Standards A - B More...
mfr. no.   USP
Featured Industry   Pharmaceutical (small molecule)
SMILES string   CC(C)c1ccc2Oc3nc(N)c(cc3C(=O)c2c1)C(O)=O
InChI   1S/C16H14N2O4/c1-7(2)8-3-4-12-9(5-8)13(19)10-6-11(16(20)21)14(17)18-15(10)22-12/h3-7H,1-2H3,(H2,17,18)(H,20,21)
InChI key   SGRYPYWGNKJSDL-UHFFFAOYSA-N
Gene Information   human ... PDE4A(5141), PDE4B(5142), PDE4C(5143), PDE4D(5144)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Amlexanox is an anti-allergy and anti-asthma drug that blocks histamine and leukotriene release from leukocytes and mast cells. Amlexanox has also been shown to inhibit cahaperone activity of Hsp90, and S100A13, which is involved in transport of proteins devoid of signal peptide sequences.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.

Sales restrictions may apply.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?