EMAIL THIS PAGE TO A FRIEND
1047503 USP

Bacitracin

United States Pharmacopeia (USP) Reference Standard

  • CAS Number 1405-87-4

  • Empirical Formula (Hill Notation) C66H103N17O16S

  • Molecular Weight 1422.69

  •  MDL number MFCD00062640

  •  PubChem Substance ID 329749095

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, USP Standards, USP Standards A - B More...
InChI Key   CLKOFPXJLQSYAH-RNHDWVCBSA-N
Featured Industry   Pharmaceutical (small molecule)

Description

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Biochem/physiol Actions

Bacitracin is a peptide antibiotic.
Antimicrobial spectrum: Gram-positive bacteria.
Mode of Action: Inhibits bacterial cell wall synthesis by inhibiting dephosphorylation of lipid pyrophosphate.

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Other Notes

Sales restrictions may apply.

USP issued SDS can be found here.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?