EMAIL THIS PAGE TO A FRIEND
1133999 USP

Allyl cinnamate

United States Pharmacopeia (USP) Reference Standard

Synonym: Allyl 3-phenylacrylate

  • CAS Number 1866-31-5

  • Linear Formula C6H5CH=CHCO2CH2CH=CH2

  • Molecular Weight 188.22

  •  MDL number MFCD00026105

  •  PubChem Substance ID 329749394

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, USP Standards, USP Standards A - B More...
grade   pharmaceutical primary standard
mfr. no.   USP
refractive index   n20/D 1.566 (lit.)
bp   150-152 °C/15 mmHg (lit.)
density   1.053 g/mL at 25 °C (lit.)
Featured Industry   Pharmaceutical (small molecule)
SMILES string   C=CCOC(=O)\C=C\c1ccccc1
InChI   1S/C12H12O2/c1-2-10-14-12(13)9-8-11-6-4-3-5-7-11/h2-9H,1,10H2/b9-8+
InChI key   KCMITHMNVLRGJU-CMDGGOBGSA-N

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?