• USA Home
  • 1220302 - Dipivefrin hydrochloride

EMAIL THIS PAGE TO A FRIEND
1220302 USP

Dipivefrin hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym: Dipivefrine hydrochloride, [2-(2,2-Dimethylpropanoyloxy)-4-[1-hydroxy-2-(methylamino)ethyl]phenyl] 2,2-dimethylpropanoate hydrochloride

  • CAS Number 64019-93-8

  • Empirical Formula (Hill Notation) C19H29NO5 · HCl

  • Molecular Weight 387.90

  •  EC Number 264-609-5

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, USP Standards, USP Standards C - D More...
Featured Industry   Pharmaceutical (small molecule)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.

Sales restrictions may apply.

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1 / PGIII

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?