EMAIL THIS PAGE TO A FRIEND
1385007 USP

Mephenytoin

United States Pharmacopeia (USP) Reference Standard

Synonym: (±)-Mephenytoin, (±)-5-Ethyl-3-methyl-5-phenyl-2,4-imidazolidinedione, (±)-5-Ethyl-3-methyl-5-phenylhydantoin

  • CAS Number 50-12-4

  • Empirical Formula (Hill Notation) C12H14N2O2

  • Molecular Weight 218.25

  •  MDL number MFCD00022407

  •  PubChem Substance ID 329750360

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, USP Standards, USP Standards L - M More...
grade   pharmaceutical primary standard
mfr. no.   USP
Featured Industry   Pharmaceutical (small molecule)
SMILES string   CCC1(NC(=O)N(C)C1=O)c2ccccc2
InChI   1S/C12H14N2O2/c1-3-12(9-7-5-4-6-8-9)10(15)14(2)11(16)13-12/h4-8H,3H2,1-2H3,(H,13,16)
InChI key   GMHKMTDVRCWUDX-UHFFFAOYSA-N
Gene Information   human ... SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN7A(6332), SCN8A(6334), SCN9A(6335)

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

CYP2B6 and CYP2C19 substrate; anticonvulsive, antiepileptic.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.

Sales restrictions may apply.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

HPLC Analysis of Mephenytoin Enantiomers on Astec® CHIROBIOTIC® V

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Mephenytoin Enantiomers on Astec® CHIROBIOTIC® V
Keywords: Anticonvulsants, Chromatography, High performance liquid chromatography, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?