EMAIL THIS PAGE TO A FRIEND
1525707 USP

Phenothiazine

United States Pharmacopeia (USP) Reference Standard

Synonym: 10H-Phenothiazine

  • CAS Number 92-84-2

  • Empirical Formula (Hill Notation) C12H9NS

  • Molecular Weight 199.27

  •  Beilstein/REAXYS Number 143237

  •  MDL number MFCD00005015

  •  PubChem Substance ID 329750802

  •  NACRES NA.24

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Pharmacopeia & Metrological Institutes Standards, USP Standards, USP Standards P - R More...
grade   pharmaceutical primary standard
mfr. no.   USP
bp   371 °C (lit.)
mp   182-187 °C (lit.)
Featured Industry   Pharmaceutical (small molecule)
SMILES string   N1c2ccccc2Sc3ccccc13
InChI   1S/C12H9NS/c1-3-7-11-9(5-1)13-10-6-2-4-8-12(10)14-11/h1-8,13H
InChI key   WJFKNYWRSNBZNX-UHFFFAOYSA-N

Description

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

USP issued SDS can be found here.

Sales restrictions may apply.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Blood
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?