• USA Home
  • V000350 - Lithium tetraborate

EMAIL THIS PAGE TO A FRIEND
V000350 Sigma-Aldrich

Lithium tetraborate

p.a., 98.0-102.0%

Purchase

Properties

Related Categories Essential Chemicals, Lithium Salts, Research Essentials, Salts
grade   p.a.
assay   98.0-102.0%
mp   760-880 °C (lit.)
density   ≥0.25 g/mL at 25 °C (lit.)
SMILES string   [Li+].[Li+].[O-]B(OB=O)OB([O-])OB=O
InChI   1S/B4O7.2Li/c5-1-9-3(7)11-4(8)10-2-6;;/q-2;2*+1
InChI key   PSHMSSXLYVAENJ-UHFFFAOYSA-N
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?