• USA Home
  • V001382 - Hydrochloric acid solution

EMAIL THIS PAGE TO A FRIEND
V001382 Sigma-Aldrich

Hydrochloric acid solution

puriss., 36%

Synonym: Hydrogen chloride solution

Purchase

Properties

vapor pressure   190 mmHg ( 25 °C)
InChI Key   VEXZGXHMUGYJMC-UHFFFAOYSA-N
grade   puriss.
assay   36%
bp   100 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1789 8 / PGII
WGK Germany 
1
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?