EMAIL THIS PAGE TO A FRIEND
V900189 Sigma-Aldrich

Sodium formate

Vetec, reagent grade, 97%

Synonym: Formic acid sodium salt

Purchase

Properties

Related Categories Essential Chemicals, Research Essentials, Salts, Sodium Salts
grade   reagent grade
product line   Vetec
assay   97%
form   chunks or granules
  crystalline powder
  powder or crystals
mp   259-262 °C (lit.)
SMILES string   [Na+].[O-]C=O
InChI   1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1
InChI key   HLBBKKJFGFRGMU-UHFFFAOYSA-M

Description

Packaging

500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?