EMAIL THIS PAGE TO A FRIEND
V900234 Sigma-Aldrich

1-Dodecanol

Vetec, reagent grade, 98%

Synonym: Alcohol C12, Dodecyl alcohol, Lauryl alcohol

Purchase

Properties

Related Categories Alcohols, Biochemicals and Reagents, Building Blocks, C9 to C10, Chemical Synthesis,
grade   reagent grade
vapor density   7.4 (vs air)
vapor pressure   0.1 mmHg ( 20 °C)
product line   Vetec
assay   98%
form   solid
autoignition temp.   500 °F
expl. lim.   4 %
refractive index   n20/D 1.442 (lit.)
bp   260-262 °C (lit.)
mp   22-26 °C (lit.)
density   0.833 g/mL at 25 °C (lit.)
SMILES string   CCCCCCCCCCCCO
InChI   1S/C12H26O/c1-2-3-4-5-6-7-8-9-10-11-12-13/h13H,2-12H2,1H3
InChI key   LQZZUXJYWNFBMV-UHFFFAOYSA-N

Description

Packaging

500 mL in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
WGK 2
Flash Point(F) 
235.4 °F - closed cup
Flash Point(C) 
113 °C - closed cup
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?