EMAIL THIS PAGE TO A FRIEND
V900297 Sigma-Aldrich

Sarcosine

Vetec reagent grade, 98%

Synonym: N-Methylglycine

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Glycine Derivatives, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
grade   Vetec reagent grade
InChI Key   FSYKKLYZXJSNPZ-UHFFFAOYSA-N
assay   98%
mp   208-212 °C (dec.) (lit.)

Description

Packaging

250 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?