EMAIL THIS PAGE TO A FRIEND
V900380 Sigma-Aldrich

MES potassium salt

Vetec reagent grade, 99%

Synonym: 2-(N-Morpholino)ethanesulfonic acid potassium salt, 4-Morpholineethanesulfonic acid potassium salt

Purchase

Properties

grade   Vetec reagent grade
assay   99%
form   crystalline powder
useful pH range   5.5 - 6.7
pKa    6.1
solubility   water: 0.333 mg/mL, clear, colorless
SMILES string   [K+].[O-]S(=O)(=O)CCN1CCOCC1
InChI   1S/C6H13NO4S.K/c8-12(9,10)6-3-7-1-4-11-5-2-7;/h1-6H2,(H,8,9,10);/q;+1/p-1
InChI key   IMFIKFZWLAWMQE-UHFFFAOYSA-M

Description

Packaging

100 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?