EMAIL THIS PAGE TO A FRIEND
V900763 Sigma-Aldrich

Anthraquinone

Vetec, reagent grade, 97%

  • CAS Number 84-65-1

  • Empirical Formula (Hill Notation) C14H8O2

  • Molecular Weight 208.21

  •  Beilstein/REAXYS Number 390030

  •  EC Number 201-549-0

  •  MDL number MFCD00001188

  •  PubChem Substance ID 329830134

Purchase

Properties

Related Categories Building Blocks, C13 to C14, Carbonyl Compounds, Chemical Synthesis, Ketones,
grade   reagent grade
vapor density   7.16 (vs air)
vapor pressure   1 mmHg ( 190 °C)
product line   Vetec
assay   97%
bp   379-381 °C (lit.)
mp   284-286 °C (lit.)
SMILES string   O=C1c2ccccc2C(=O)c3ccccc13
InChI   1S/C14H8O2/c15-13-9-5-1-2-6-10(9)14(16)12-8-4-3-7-11(12)13/h1-8H
InChI key   RZVHIXYEVGDQDX-UHFFFAOYSA-N

Description

Packaging

100, 500 g in poly bottle

Legal Information

Vetec is a trademark of Sigma-Aldrich International GmbH

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
365.0 °F - closed cup
Flash Point(C) 
185 °C - closed cup
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?