• USA Home
  • Search Results

  • 10hydroxydecanoicacid18826167953411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

10-Hydroxydecanoic acid

1 Products
Synonym:   NSC 15139
Image of 10-Hydroxydecanoic acid
CAS Number:
Linear Formula:
 HO(CH2)9CO2H
Molecular Weight:
 188.26
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More