• USA Home
  • Search Results

  • 110phenanthrolinemonohydrate19822514489811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,10-Phenanthroline monohydrate

4 Products
Synonym:   o-Phenanthroline monohydrate
Image of 1,10-Phenanthroline monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C12H8N2 · H2O
Molecular Weight:
 198.22
 Beilstein/REAXYS Number:
 4008096
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
  Physical Form
  Melting Point
  Suitability
  Application(s)
  Special Grade
Show More