• USA Home
  • Search Results

  • 11122pentafluoro4iodobutane273974072380611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1,1,2,2-Pentafluoro-4-iodobutane

1 Products
Synonym:   1H,1H,2H,2H-Perfluorobutyl iodide
Image of 1,1,1,2,2-<WBR>Pentafluoro-<WBR>4-<WBR>iodobutane
CAS Number:
Empirical Formula (Hill Notation):
 C4H4F5I
Molecular Weight:
 273.97
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More