• USA Home
  • Search Results

  • 1112tetrafluoroethane1020381197211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

1,1,1,2-Tetrafluoroethane

1 Products
Synonym:   HFC-134a
Image of 1,1,1,2-<WBR>Tetrafluoroethane
CAS Number:
Linear Formula:
 FCH2CF3
Molecular Weight:
 102.03
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More